missing translation for 'onlineSavingsMsg'
Learn More

Fibrinopeptide B Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™

Product Code. 30498993 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity unitSize
30498993 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy

Product Code. 30498993

Brand: Novus Biologicals NBP335409AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fibrinopeptide B Polyclonal antibody specifically detects Fibrinopeptide B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrinopeptide B
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias fibrinogen beta chain, fibrinogen, B beta polypeptide, MGC104327, MGC120405
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 111-222 of human Fibrinopeptide B (NP_005132.2).,, Sequence:, QLQEALLQQERPIRNSVDELNNNVEAVSQTSSSSFQYMYLLKDLWQKRQKQVKDNENVVNEYSSELEKHQLYIDETVNSNIPTNLRVLRSILENLRSKIQKLESDVSAQMEY
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2244
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.