missing translation for 'onlineSavingsMsg'
Learn More

HIBADH Antibody [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30498965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity unitSize
30498965 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy

Product Code. 30498965

Brand: Novus Biologicals NBP337923UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIBADH Polyclonal antibody specifically detects HIBADH in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIBADH
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias 3-hydroxyisobutyrate dehydrogenase, 3-hydroxyisobutyrate dehydrogenase, mitochondrial, EC 1.1.1,3'-hydroxyisobutyrate dehydrogenase, EC 1.1.1.31, MGC40361, NS5ATP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 37-336 of human HIBADH (NP_689953.1).,, Sequence:, ASKTPVGFIGLGNMGNPMAKNLMKHGYPLIIYDVFPDACKEFQDAGEQVVSSPADVAEKADRIITMLPTSINAIEAYSGANGILKKVKKGSLLIDSSTIDPAVSKELAKEVEKMGAVFMDAPVSGGVGAARSGNLTFMVGGVEDEFAAAQELLGCMGSNVVYCGAVGTGQAAKICNNMLLAISMIGTAEAMNLGIRLGLDPKLLAKILNMSSGRCWSSDTYNPVPGVMDGVPSANNYQGGFGTTLMAKDLGLAQDSATSTKSPILLGSLAHQIYRMMCAKGYSKKDFSSVFQFLREEETF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11112
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.