missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S beta2 Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30498942 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498942 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498942

Brand: Novus Biologicals NBP335371AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Proteasome 20S beta2 Polyclonal antibody specifically detects Proteasome 20S beta2 in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proteasome 20S beta2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.25.1, HC7-I, Macropain subunit C7-I, MGC104215, MGC126885, multicatalytic endopeptidase complex subunit C7-1, Multicatalytic endopeptidase complex subunit C7-I, proteasome (prosome, macropain) subunit, beta type, 2, proteasome beta 2 subunit, Proteasome component C7-I, proteasome subunit beta type-2, proteasome subunit, beta type, 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human Proteasome 20S beta2 (NP_002785.1).,, Sequence:, MEYLIGIQGPDYVLVASDRVAASNIVQMKDDHDKMFKMSEKILLLCVGEAGDTVQFAEYIQKNVQLYKMRNGYELSPTAAANFTRRNLADCLRSRTPYHVNLLLAGYDEHEGPALYYMDYLAALAKAPFAAHGYGAFLTLSILDRYYTPTISRERAVELLRKCLEELQKRFILNLPTFSVRIIDKNGIHDLDNISFPKQGS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 5690
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.