missing translation for 'onlineSavingsMsg'
Learn More

TEAD4 Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30498914 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Product Code. Quantity
30498914 0.1 mL
1 options
This item is not returnable. View return policy

Product Code. 30498914

Brand: Novus Biologicals NBP338077FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TEAD4 Polyclonal antibody specifically detects TEAD4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEAD4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EFTR-2, hRTEF-1B, related transcription enhancer factor 1B, RTEF1RTEF-1, TCF13L1MGC9014, TEA domain family member 4TEF3, TEAD-4, TEF-3, TEFR-1, Transcription factor 13-like 1, Transcription factor RTEF-1, transcriptional enhancer factor 1-related, transcriptional enhancer factor 3, transcriptional enhancer factor TEF-3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-250 of human TEAD4 (NP_003204.2).,, Sequence:, AREIQAKLKDQAAKDKALQSMAAMSSAQIISATAFHSSMALARGPGRPAVSGFWQGALPGQAGTSHDVKPFSQQTYAVQPPLPLPGFESPAGPAPSPSAPPAPPWQGRSVASSKLWMLEFSAFLEQQQDPDTYNKHLFVHI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7004
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.