missing translation for 'onlineSavingsMsg'
Learn More

EPLIN Antibody [Biotin], Novus Biologicals Biologicals™

Product Code. 30498892 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498892 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498892

Brand: Novus Biologicals NBP327896B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EPLIN Polyclonal antibody specifically detects EPLIN in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPLIN
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias 1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3
Host Species Rabbit
Immunogen EPLIN antibody was raised against an 18 amino acid peptide near the amino terminus of human EPLIN. The immunogen is located within amino acids 230 - 280 of EPLIN.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51474
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.