missing translation for 'onlineSavingsMsg'
Learn More

CINP Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30498885 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498885 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498885

Brand: Novus Biologicals NBP337914AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CINP Polyclonal antibody specifically detects CINP in Human,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CINP
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human CINP (NP_116019.1).,, Sequence:, MEAKTLGTVTPRKPVLSVSARKIKDNAADWHNLILKWETLNDAGFTTANNIANLKISLLNKDKIELDSSSPASKENEEKVCLEYNEELEKLCEELQATLDGLTKIQVKMEKLSSTTKGICELENYHYGEESKRPPLFHTWPTTHFYEVSHKLLEMYRKELLLKRTVAKELAHTGDPDLTLSYLSMWLHQPYVESDSRLHLESMLLETGHRAL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51550
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.