missing translation for 'onlineSavingsMsg'
Learn More

Spike Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™

Product Code. 30498884 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498884 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498884

Brand: Novus Biologicals NBP337899AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Spike Polyclonal antibody specifically detects Spike in SARS-CoV samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spike
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 600-700 of coronavirus Spike (NP_828851.1).,, Sequence:, DVNCTDVSTAIHADQLTPAWRIYSTGNNVFQTQAGCLIGAEHVDTSYECDIPIGAGICASYHTVSLLRSTSQKSIVAYTMSLGADSSIAYSNNTIAIPTNF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.