missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 2 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Cytokeratin 2 Polyclonal antibody specifically detects Cytokeratin 2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Cytokeratin 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CK-2e, Cytokeratin-2e, epidermal ichthyosis bullosa of Siemens, Epithelial keratin-2e, K2e, keratin 2, keratin 2A (epidermal ichthyosis bullosa of Siemens), keratin, type II cytoskeletal 2 epidermal, Keratin-2 epidermis, keratin-2e, KRT2AMGC116967, KRT2EMGC116968, KRTE, Type-II keratin Kb2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human Cytokeratin 2 (KRT2) (NP_000414.2).,, Sequence:, MSCQISCKSRGRGGGGGGFRGFSSGSAVVSGGSRRSTSSFSCLSRHGGGGGGFGGGGFGSRSLVGLGGTKSISISVAGGGGGFGAAGGFGGRGGGFGGGSSFGGGSGFSGGGFGGGGFGGGRFGGFGGPGGVGGLGGPGGFGPGGYPGGIHEVSVNQSLLQPLNVKVDPEIQNVKAQEREQIKTLNNKFASFIDKVRFLEQQNQVLQTKWELLQQMNVGT |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?