missing translation for 'onlineSavingsMsg'
Learn More

Methionine Sulfoxide Reductase A Antibody [Biotin], Novus Biologicals Biologicals™

Product Code. 30498776 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498776 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498776

Brand: Novus Biologicals NBP336719B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Methionine Sulfoxide Reductase A Polyclonal antibody specifically detects Methionine Sulfoxide Reductase A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methionine Sulfoxide Reductase A
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias cytosolic methionine-S-sulfoxide reductase, EC 1.8.4.11, methionine sulfoxide reductase A, peptide met (O) reductase, Peptide Met(O) reductase, peptide methionine sulfoxide reductase, Peptide-methionine (S)-S-oxide reductase, PMSR, Protein-methionine-S-oxide reductase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-235 of human Methionine Sulfoxide Reductase A (NP_036463.1).,, Sequence:, GNSASNIVSPQEALPGRKEQTPVAAKHHVNGNRTVEPFPEGTQMAVFGMGCFWGAERKFWVLKGVYSTQVGFAGGYTSNPTYKEVCSEKTGHAEVVRVVYQPEHMSFEELLKVFWENHDPTQGMRQGNDHGTQYRSAIYPTSAKQMEAALSSKENYQKVLSEHGFGPITTDIREGQTFYYAEDYHQQYLSKNPNGYCGLGGTGVSCPVGIKK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4482
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.