missing translation for 'onlineSavingsMsg'
Learn More

TRIM47 Antibody [Biotin], Novus Biologicals Biologicals™

Product Code. 30498715 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498715 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498715

Brand: Novus Biologicals NBP335790B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM47 Polyclonal antibody specifically detects TRIM47 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM47
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Gene overexpressed in astrocytoma protein, GOARING finger protein 100, RNF100tripartite motif-containing protein 47, tripartite motif containing 47, tripartite motif-containing 47
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 356-530 of human TRIM47 (NP_258411.2).,, Sequence:, ELSFTKSSQAVRAVRDMLAVACVNQWEQLRGPGGNEDGPQKLDSEADAEPQDLESTNLLESEAPRDYFLKFAYIVDLDSDTADKFLQLFGTKGVKRVLCPINYPLSPTRFTHCEQVLGEGALDRGTYYWEVEIIEGWVSMGVMAEDFSPQEPYDRGRLGRNAHSCCLQWNGRSFS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 91107
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.