missing translation for 'onlineSavingsMsg'
Learn More

Caspase-12 Antibody [Alexa Fluor« 350], R&D Systems™

Product Code. 30498668 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize':
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30498668 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30498668 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP335067AF350
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Caspase-12 Polyclonal antibody specifically detects Caspase-12 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-12
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Caspase-12 (NP_001177945.2).,, Sequence:, FNNRNCQSLKDKPKVIIMQACRGNGAGIVWFTTDSGKASADTHGRLLQGNICNDAVTKAHVEKDFIAFKSSTPHNVSWRHETNGSVFISQIIYYFREYSWS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 100506742
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.