missing translation for 'onlineSavingsMsg'
Learn More

POLR2E Antibody [DyLight 594], R&D Systems™

Product Code. 30498634 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30498634 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498634 Supplier Novus Biologicals Supplier No. NBP335759DL594
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

POLR2E Polyclonal antibody specifically detects POLR2E in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR2E
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DNA directed RNA polymerase II 23 kda polypeptide, DNA-directed RNA polymerase II 23 kDa polypeptide, DNA-directed RNA polymerase II subunit E, DNA-directed RNA polymerases I, II, and III subunit RPABC1, hRPB25, hsRPB5, polymerase (RNA) II (DNA directed) polypeptide E (25kD), polymerase (RNA) II (DNA directed) polypeptide E, 25kDa, RPABC1, RPB5, RPB5 homolog, XAP4RNA polymerases I, II, and III subunit ABC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human POLR2E (NP_002686.2).,, Sequence:, MDDEEETYRLWKIRKTIMQLCHDRGYLVTQDELDQTLEEFKAQFGDKPSEGRPRRTDLTVLVAHNDDPTDQMFVFFPEEPKVGIKTIKVYCQRMQEENITRALIVVQQGMTPSAKQSLVDMAPKYILEQFLQQELLINITEHELVPEHVVMTKEEVTELLARYKLRENQLPRIQAGDPVARYFGIKRGQVVKIIRPSETAGRYITYRLVQ
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5434
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.