missing translation for 'onlineSavingsMsg'
Learn More

BRAF35 Antibody [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30498620 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30498620 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498620 Supplier Novus Biologicals Supplier No. NBP337920R
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRAF35 Polyclonal antibody specifically detects BRAF35 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRAF35
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias BRAF35Sox-like transcriptional factor, high-mobility group 20B, HMG domain-containing protein 2, HMG domain-containing protein HMGX2, HMGX2FLJ26127, HMGXB2member 1-related, SMARCE1R, SMARCE1-related protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human BRAF35 (NP_006330.2).,, Sequence:, MSHGPKQPGAAAAPAGGKAPGQHGGFVVTVKQERGEGPRAGEKGSHEEEPVKKRGWPKGKKRKKILPNGPKAPVTGYVRFLNERREQIRTRHPDLPFPEITKMLGAEWSKLQPTEKQRYLDEAEREKQQYMKELRAYQQSEAYKMCTEKIQEKKIKKEDSSSGLMNTLLNGHKGGDCDGFSTFDVPIFTEEFLDQNKAREAELRRLRKMNVAFEEQNAVLQRHTQSMSSARERLEQELALEERRTLALQQQLQAVRQALTASFASLPVPGTGETPTLGTLDFYMARLHGAIERDPAQHEKLIVRIKEILAQVASEHL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10362
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.