missing translation for 'onlineSavingsMsg'
Learn More

ACSL3 Antibody [PerCP], R&D Systems™

Product Code. 30498589 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30498589 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498589 Supplier Novus Biologicals Supplier No. NBP335183PCP
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ACSL3 Polyclonal antibody specifically detects ACSL3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL3
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias ACS3EC 6.2.1.3, acyl-CoA synthetase long-chain family member 3, FACL3lignoceroyl-CoA synthase, fatty-acid-Coenzyme A ligase, long-chain 3, LACS 3, LACS3, Long-chain acyl-CoA synthetase 3, long-chain-fatty-acid--CoA ligase 3, PRO2194
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 621-720 of human ACSL3 (NP_004448.2).,, Sequence:, VIGFVVPNQKELTELARKKGLKGTWEELCNSCEMENEVLKVLSEAAISASLEKFEIPVKIRLSPEPWTPETGLVTDAFKLKRKELKTHYQADIERMYGRK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2181
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.