missing translation for 'onlineSavingsMsg'
Learn More

alcohol dehydrogenase 5 Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30498580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498580 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498580 Supplier Novus Biologicals Supplier No. NBP335343CL1
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

alcohol dehydrogenase 5 Polyclonal antibody specifically detects alcohol dehydrogenase 5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen alcohol dehydrogenase 5
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias ADH-3, ADHXEC 1.1.1.1, alcohol dehydrogenase (class III), chi polypeptide, Alcohol dehydrogenase 5, alcohol dehydrogenase 5 (class III), chi polypeptide, Alcohol dehydrogenase class chi chain, alcohol dehydrogenase class-3, Alcohol dehydrogenase class-III, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.284, FALDH, FDHGSNOR, formaldehyde dehydrogenase, Glutathione-dependent formaldehyde dehydrogenase, GSH-FDH, S-(hydroxymethyl)glutathione dehydrogenase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-374 of human alcohol dehydrogenase 5 (NP_000662.3).,, Sequence:, MANEVIKCKAAVAWEAGKPLSIEEIEVAPPKAHEVRIKIIATAVCHTDAYTLSGADPEGCFPVILGHEGAGIVESVGEGVTKLKAGDTVIPLYIPQCGECKFCLNPKTNLCQKIRVTQGKGLMPDGTSRFTCKGKTILHYMGTSTFSEYTVVADISVAKIDPLAPLDKVCLLGCGISTGYGAAVNTAKLEPGSVCAVFGLGGVGLAVIMGCKVAGASRIIGVDINKDKFARAKEFGATECINPQDFSKPIQEVLIEMTDGGVDYSFECIGNVKVMRAALEACHKGWGVSVVVGVAASGEEIATRPFQLVTGRTWKGTAFGGWKSVESVPKLVSEYMSKKIKVDEFVTHNLSFDEINKAFELMHSGKSIRTVVKI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 128
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.