missing translation for 'onlineSavingsMsg'
Learn More

ITPKB Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30498574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498574 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498574

Brand: Novus Biologicals NBP338060FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ITPKB Polyclonal antibody specifically detects ITPKB in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPKB
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.1.127, inositol 14,5-trisphosphate 3-kinase BIP3K, inositol-trisphosphate 3-kinase B, InsP 3-kinase B, IP3 3-kinase B, IP3-3KB, IP3K B, IP3KB, IP3K-B, PIG37, proliferation-inducing protein 37
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ITPKB (NP_002212.3).,, Sequence:, MAVYCYALNSLVIMNSANEMKSGGGPGPSGSETPPPPRRAVLSPGSVFSPGRGASFLFPPAESLSPEEPRSPGGWRSGRRRLNSSSGSGSGSSGSSVSSPSWAGRLRGDRQQVVAAGTLSPPGPEEAKRKLRILQRELQNVQVNQKVGMFEAHIQAQSSAIQAPRSPRLGRARSPSPCPFRSSSQPPGRVLVQGARSEERRTKSWGEQCPETSGTDSGRKGGPSLCSSQVKKGMPPLPGRAAPTGSEAQGPSAFVRMEKG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3707
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.