missing translation for 'onlineSavingsMsg'
Learn More

H2A Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30498561 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498561 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498561

Brand: Novus Biologicals NBP335820C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

H2A Polyclonal antibody specifically detects H2A in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias H2A histone family, member O, H2A.2, H2A/O, H2A/q, H2a-615, H2AFOHistone H2A/o, HIST2H2AAH2A, histone 2, H2aa, histone 2, H2aa3, histone cluster 2, H2aa3, histone H2A type 2-A, Histone H2A.2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-208 of mouse H2A (NP_034508.2).,, Sequence:, IEADHVGTYGISVYQSPGDIGQYTFEFDGDELFYVDLDKKETVWMLPEFGQLASFDPQGGLQNIAVVKHNLGVLTKRSNSTPATNEAPQATVFPKSPVLLGQPNTLICFVDNIFPPVINITWLRNSKSVADGVYETSFFVNRDYSFHKLSYLTFIPSDDDIYDCKVEHWGLEEPVLKHWEPE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8337
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.