missing translation for 'onlineSavingsMsg'
Learn More

Smad1 Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30498554 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498554 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498554

Brand: Novus Biologicals NBP335165APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Smad1 Polyclonal antibody specifically detects Smad1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smad1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias BSP1, BSP-1, hSMAD1, JV4-1SMAD, mothers against DPP homolog 1 (Drosophila), MAD homolog 1, MADH1JV41, MADR1MAD, mothers against decapentaplegic homolog 1 (Drosophila), Mad-related protein 1, mothers against decapentaplegic homolog 1, Mothers against DPP homolog 1, SMAD family member 1SMAD 1, SMAD, mothers against DPP homolog 1, Smad1, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-240 of human SMAD1/5/8/9 (NP_001003688.1).,, Sequence:, WKQGDEEEKWAEKAVDALVKKLKKKKGAMEELEKALSCPGQPSNCVTIPRSLDGRLQVSHRKGLPHVIYCRVWRWPDLQSHHELKPLECCEFPFGSKQKEVCINPYHYKRVESPVLPPVLVPRHSEYNPQHSLLAQFRNLGQNEPHMPLNATFPDSFQQPNSHPFPHSPNSSYPNSPGSSSSTYPHSPTSSDPGSPFQMPADTPPPAYLPPEDPMTQDGSQ
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4086
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.