missing translation for 'onlineSavingsMsg'
Learn More

Laminin beta 3 Antibody [Biotin], Novus Biologicals Biologicals™

Product Code. 30498523 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498523 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498523 Supplier Novus Biologicals Supplier No. NBP335175B
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Laminin beta 3 Polyclonal antibody specifically detects Laminin beta 3 in Human,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 762-1004 of human Laminin beta 3 (NP_000219.2).,, Sequence:, QQACSRGACYPPVGDLLVGRTRFLRASSTCGLTKPETYCTQYGEWQMKCCKCDSRQPHNYYSHRVENVASSSGPMRWWQSQNDVNPVSLQLDLDRRFQLQEVMMEFQGPMPAGMLIERSSDFGKTWRVYQYLAADCTSTFPRVRQGRPQSWQDVRCQSLPQRPNARLNGGKVQLNLMDLVSGIPATQSQKIQEVGEITNLRVNFTRLAPVPQR
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3914
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.