missing translation for 'onlineSavingsMsg'
Learn More

XRCC6BP1 Antibody [Alexa Fluor« 350], Novus Biologicals Biologicals™

Product Code. 30498507 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30498507 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498507 Supplier Novus Biologicals Supplier No. NBP335282AF350
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XRCC6BP1 Polyclonal antibody specifically detects XRCC6BP1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRCC6BP1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.-, Ku70-binding protein 3, KUB3Ku70 binding protein 3, MGC134817, MGC134818, mitochondrial inner membrane protease ATP23 homolog, XRCC6 binding protein 1, XRCC6-binding protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-246 of human XRCC6BP1 (NP_150592.1).,, Sequence:, MAGAPDERRRGPAAGEQLQQQHVSCQVFPERLAQGNPQQGFFSSFFTSNQKCQLRLLKTLETNPYVKLLLDAMKHSGCAVNKDRHFSCEDCNGNVSGGFDASTSQIVLCQNNIHNQAHMNRVVTHELIHAFDHCRAHVDWFTNIRHLACSEVRAANLSGDCSLVNEIFRLHFGLKQHHQTCVRDRATLSILAVRNISKEVAKKAVDEVFESCFNDHEPFGRIPHNKTYARYAHRDFENRDRYYSNI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 91419
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.