missing translation for 'onlineSavingsMsg'
Learn More

VARS Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30498497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Product Code. Quantity
30498497 0.1 mL
1 options
This item is not returnable. View return policy

Product Code. 30498497

Brand: Novus Biologicals NBP338079C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VARS Polyclonal antibody specifically detects VARS in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen VARS
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias EC 6.1.1, EC 6.1.1.9, G7AVARS1, Protein G7a, valine tRNA ligase 1, cytoplasmic, Valine--tRNA ligase, ValRS, valyl-tRNA synthetase, valyl-tRNA synthetase 2, VARS2valRS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human VARS (NP_006286.1).,, Sequence:, MSTLYVSPHPDAFPSLRALIAARYGEAGEGPGWGGAHPRICLQPPPTSRTPFPPPRLPALEQGPGGLWVWGATAVAQLLWPAGLGGPGGSRAAVLVQQWVSYADTELIPAACGATLPALGLRSSAQDPQAVLGALGRALSPLEEWLRLHTYLAGEAPTLADLAAVTALLLPFRYVLDPPARRIWNNVTRWFVTCVRQPEFRAVLGEVVLYSGARPLSHQPGPEAPALPKTAAQLKKEAKKREKLEKFQQKQKIQQQQPPPGEKKPKPEKREKRDPGVITY
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7407
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.