missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
APEH Polyclonal antibody specifically detects APEH in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | APEH |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AARE, ACPH, acylamino-acid-releasing enzyme, acylaminoacyl-peptidase, Acyl-peptide hydrolase, APH, D3F15S2, D3S48EEC 3.4.19.1, DNF15S2, MGC2178, N-acylaminoacyl-peptide hydrolase, OPH, Oxidized protein hydrolase |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human APEH (NP_001631.3).,, Sequence:, MERQVLLSEPEEAAALYRGLSRQPALSAACLGPEVTTQYGGQYRTVHTEWTQRDLERMENIRFCRQYLVFHDGDSVVFAGPAGNSVETRGELLSRESPSGTMKAVLRKAGGTGPGEEKQFLEVWEKNRKLKSFNLSALEKHGPVYEDDCFGCLSWSHSETHLLYVAEKKRPKAESFFQTKALDVSASDDEIARLKKPDQAIKGDQFVFYEDWGENMVSKSIPVLCVLDVESGNISVLEGVPENVSPGQAFWAPGDAGVVF |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?