missing translation for 'onlineSavingsMsg'
Learn More

METT10D Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30498386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498386 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498386

Brand: Novus Biologicals NBP335618AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

METT10D Polyclonal antibody specifically detects METT10D in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen METT10D
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.-, methyltransferase 10 domain containing, Methyltransferase 10 domain-containing protein, methyltransferase like 16, METT10D, MGC3329, putative methyltransferase METT10D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 263-562 of human METT10D (NP_076991.3).,, Sequence:, RIQGVPKVTYTEFCQGRTMRWALAWSFYDDVTVPSPPSKRRKLEKPRKPITFVVLASVMKELSLKASPLRSETAEGIVVVTTWIEKILTDLKVQHKRVPCGKEEVSLFLTAIENSWIHLRRKKRERVRQLREVPRAPEDVIQALEEKKPTPKESGNSQELARGPQERTPCGPALREGEAAAVEGPCPSQESLSQEENPEPTEDERSEEKGGVEVLESCQGSSNGAQDQEASEQFGSPVAERGKRLPGVAGQYLFKCLINVKKEVDDALVEMHWVEGQNRDLMNQLCTYIRNQIFRLVAVN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79066
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.