missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BTF Polyclonal specifically detects BTF in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Antigen | BTF |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1 |
| Host Species | Rabbit |
| Immunogen | BTF antibody was raised against a 19 amino acid peptide near the carboxy terminus of human BTF. The immunogen is located within amino acids 820 - 870 of BTF. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?