missing translation for 'onlineSavingsMsg'
Learn More

PAWR/ PAR4 Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30498354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498354 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498354 Supplier Novus Biologicals Supplier No. NBP338026MFV610
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PAWR / PAR4 Polyclonal antibody specifically detects PAWR / PAR4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAWR / PAR4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Par-4, PAR4prostate apoptosis response protein 4, PRKC apoptosis WT1 regulator protein, PRKC, apoptosis, WT1, regulator, Prostate apoptosis response 4 protein, prostate apoptosis response protein PAR-4, transcriptional repressor PAR4, WT1-interacting protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human PAWR/ PAR4 (NP_002574.2),, Sequence:, QNEAVNLLDPGSSYLLQEPPRTVSGRYKSTTSVSEEDVSSRYSRTDRSGFPRYNRDANVSGTLVSSSTLEKKIEDLEKEVVRERQENLRLVRLMQDKEEMI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5074
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.