missing translation for 'onlineSavingsMsg'
Learn More

FANCI Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30498319 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498319 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498319 Supplier Novus Biologicals Supplier No. NBP338040V
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FANCI Polyclonal antibody specifically detects FANCI in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCI
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1061-1328 of human FANCI (NP_001106849.1).,, Sequence:, DQDVEVEKTNHFAIVNLRTAAPTVCLLVLSQAEKVLEEVDWLITKLKGQVSQETLSEEASSQATLPNQPVEKAIIMQLGTLLTFFHELVQTALPSGSCVDTLLKDLCKMYTTLTALVRYYLQVCQSSGGIPKNMEKLVKLSGSHLTPLCYSFISYVQNKSKSLNYTGEKKEKPAAVATAMARVLRETKPIPNLIFAIEQYEKFLIHLSKKSKVNLMQHMKLSTSRDFKIKGNILDMVLREDGEDENEEGTASEHGGQNKEPAKKKRKK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55215
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.