missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAAH Polyclonal antibody specifically detects FAAH in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | FAAH |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Anandamide amidohydrolase 1, EC 3.5.1, EC 3.5.1.99, EC 3.5.1.n2, FAAH1, FAAH-1, fatty acid amide hydrolase, fatty-acid amide hydrolase 1, MGC102823, MGC138146, Oleamide hydrolase 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 450-579 of human FAAH (NP_001432.2).,, Sequence:, EIEVYRKTVIAQWRALDLDVVLTPMLAPALDLNAPGRATGAVSYTMLYNCLDFPAGVVPVTTVTAEDEAQMEHYRGYFGDIWDKMLQKGMKKSVGLPVAVQCVALPWQEELCLRFMREVERLMTPEKQSS |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?