missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoleucyl tRNA synthetase Antibody [DyLight 680], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Isoleucyl tRNA synthetase Polyclonal antibody specifically detects Isoleucyl tRNA synthetase in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Isoleucyl tRNA synthetase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 6.1.1, EC 6.1.1.5, FLJ20736, IARS1, ILERS, ILRS, IRS, isoleucine tRNA ligase 1, cytoplasmic, Isoleucine--tRNA ligase, isoleucyl-tRNA synthetase, isoleucyl-tRNA synthetase, cytoplasmic, PRO0785 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1158-1262 of human Isoleucyl tRNA synthetase (NP_002152.2).,, Sequence:, SAPSLINSSSTLLCQYINLQLLNAKPQECLMGTVGTLLLENPLGQNGLTHQGLLYEAAKVFGLRSRKLKLFLNETQTQEITEDIPVKTLNMKTVYVSVLPTTADF |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?