missing translation for 'onlineSavingsMsg'
Learn More

INPP5A Antibody [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30498244 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498244 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498244

Brand: Novus Biologicals NBP338035IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

INPP5A Polyclonal antibody specifically detects INPP5A in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 43 kDa inositol polyphosphate 5-phophatase, CTCL tumor antigen HD-CL-02,5PTASE, DKFZp434A1721, EC 3.1.3.56, inositol polyphosphate-5-phosphatase, 40kD, inositol polyphosphate-5-phosphatase, 40kDa, inositol trisphosphate-5-phosphatase, 40kD, InsP3 5-phosphatase, MGC116947,5PTase, MGC116949, type I inositol-14,5-trisphosphate 5-phophatase, type I inositol-14,5-trisphosphate 5-phosphatase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 10-200 of human INPP5A (NP_005530.3).,, Sequence:, TAVLLVTANVGSLFDDPENLQKNWLREFYQVVHTHKPHFMALHCQEFGGKNYEASMSHVDKFVKELLSSDAMKEYNRARVYLDENYKSQEHFTALGSFYFLHESLKNIYQFDFKAKKYRKVAGKEIYSDTLESTPMLEKEKFPQDYFPECKWSRKGFIRTRWCIADCAFDLVNIHLFHDASNLVAWETSPS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 3632
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.