missing translation for 'onlineSavingsMsg'
Learn More

ETEA Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™

Product Code. 30498241 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498241 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498241

Brand: Novus Biologicals NBP337892AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ETEA Polyclonal antibody specifically detects ETEA in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETEA
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias B17.2L, ETEAFAS-associated factor 2, expressed in T-cells and eosinophils in atopic dermatitis, Fas associated factor family member 2, KIAA0887UBX domain protein 3B, MMTN, NDUNDUFA12L, UBX domain containing 8, UBX domain-containing protein 3B, UBX domain-containing protein 8, UBXD8, UBXN3BProtein ETEA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 176-393 of human ETEA (NP_055428.1).,, Sequence:, YLHGDDHQDSDEFCRNTLCAPEVISLINTRMLFWACSTNKPEGYRVSQALRENTYPFLAMIMLKDRRMTVVGRLEGLIQPDDLINQLTFIMDANQTYLVSERLEREERNQTQVLRQQQDEAYLASLRADQEKERKKREERERKRRKEEEVQQQKLAEERRRQNLQEEKERKLECLPPEPSPDDPESVKIIFKLPNDSRVERRFHFSQSLTVIHDFLFS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23197
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.