missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CYP51A1 Polyclonal antibody specifically detects CYP51A1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | CYP51A1 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CP51, CYP51Cytochrome P450LI, CYPL1, CYPLI, Cytochrome P450 51A1, cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, EC 1.14.13.70, lanosterol 14-alpha demethylase, lanosterol 14-alpha-demethylase, LDMCytochrome P45014DM, P450-14DM, P450L1,51 (lanosterol 14-alpha-demethylase), Sterol 14-alpha demethylase |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 245-420 of CYP51A1 (NP_000777.1).,, Sequence:, WLLPGWLPLPSFRRRDRAHREIKDIFYKAIQKRRQSQEKIDDILQTLLDATYKDGRPLTDDEVAGMLIGLLLAGQHTSSTTSAWMGFFLARDKTLQKKCYLEQKTVCGENLPPLTYDQLKDLNLLDRCIKETLRLRPPIMIMMRMARTPQTVAGYTIPPGHQVCVSPTVNQRLKDS |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?