missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Potassium ATPase Beta Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30498184 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498184 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498184

Brand: Novus Biologicals NBP335098JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hydrogen Potassium ATPase Beta Polyclonal antibody specifically detects Hydrogen Potassium ATPase Beta in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hydrogen Potassium ATPase Beta
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 62-291 of human Hydrogen Potassium ATPase Beta (NP_000696.1).,, Sequence:, LMQTVDPYTPDYQDQLRSPGVTLRPDVYGEKGLEIVYNVSDNRTWADLTQTLHAFLAGYSPAAQEDSINCTSEQYFFQESFRAPNHTKFSCKFTADMLQNCSGLADPNFGFEEGKPCFIIKMNRIVKFLPSNGSAPRVDCAFLDQPRELGQPLQVKYYPPNGTFSLHYFPYYGKKAQPHYSNPLVAAKLLNIPRNAEVAIVCKVMAEHVTFNNPHDPYEGKVEFKLKIEK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 496
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.