missing translation for 'onlineSavingsMsg'
Learn More

P4HA1 Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30498156 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498156 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498156

Brand: Novus Biologicals NBP335149FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

P4HA1 Polyclonal antibody specifically detects P4HA1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen P4HA1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), 2-oxoglutarate-4-dioxygenase subunit alpha-1, 4-PH alpha-1, alpha polypeptide I, EC 1.14.11.2, Procollagen-proline, prolyl 4-hydroxylase, alpha polypeptide I
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 235-534 of human P4HA1 (NP_001017962.1).,, Sequence:, DPEHQRANGNLKYFEYIMAKEKDVNKSASDDQSDQKTTPKKKGVAVDYLPERQKYEMLCRGEGIKMTPRRQKKLFCRYHDGNRNPKFILAPAKQEDEWDKPRIIRFHDIISDAEIEIVKDLAKPRLRRATISNPITGDLETVHYRISKSAWLSGYENPVVSRINMRIQDLTGLDVSTAEELQVANYGVGGQYEPHFDFARKDEPDAFKELGTGNRIATWLFYMSDVSAGGATVFPEVGASVWPKKGTAVFWYNLFASGEGDYSTRHAACPVLVGNKWVSNKWLHERGQEFRRPCTLSELE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5033
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.