missing translation for 'onlineSavingsMsg'
Learn More

TRIB1 Antibody [DyLight 755], Novus Biologicals Biologicals™

Product Code. 30498123 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498123 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498123

Brand: Novus Biologicals NBP335100IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIB1 Polyclonal antibody specifically detects TRIB1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIB1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias C8FWG-protein-coupled receptor induced protein, GIG-2, GIG2G-protein-coupled receptor-induced protein 2, G-protein-coupled receptor-induced gene 2 protein, SKIP1, TRB-1, TRB1phosphoprotein regulated by mitogenic pathways, tribbles homolog 1, tribbles homolog 1 (Drosophila), tribbles-like protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 203-372 of human TRIB1 (NP_079471.1).,, Sequence:, LGDLKLRKFVFSTEERTQLRLESLEDTHIMKGEDDALSDKHGCPAYVSPEILNTTGTYSGKAADVWSLGVMLYTLLVGRYPFHDSDPSALFSKIRRGQFCIPEHISPKARCLIRSLLRREPSERLTAPEILLHPWFESVLEPGYIDSEIGTSDQIVPEYQEDSDISSFFC
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10221
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.