missing translation for 'onlineSavingsMsg'
Learn More

FANCI Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30498073 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498073 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498073

Brand: Novus Biologicals NBP338040JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FANCI Polyclonal antibody specifically detects FANCI in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCI
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1061-1328 of human FANCI (NP_001106849.1).,, Sequence:, DQDVEVEKTNHFAIVNLRTAAPTVCLLVLSQAEKVLEEVDWLITKLKGQVSQETLSEEASSQATLPNQPVEKAIIMQLGTLLTFFHELVQTALPSGSCVDTLLKDLCKMYTTLTALVRYYLQVCQSSGGIPKNMEKLVKLSGSHLTPLCYSFISYVQNKSKSLNYTGEKKEKPAAVATAMARVLRETKPIPNLIFAIEQYEKFLIHLSKKSKVNLMQHMKLSTSRDFKIKGNILDMVLREDGEDENEEGTASEHGGQNKEPAKKKRKK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55215
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.