missing translation for 'onlineSavingsMsg'
Learn More

ATIC Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30498040 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498040 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498040

Brand: Novus Biologicals NBP337925JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATIC Polyclonal antibody specifically detects ATIC in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATIC
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase, 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase, AICAR, AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme, AICARFT, AICARFT/IMPCHASE, bifunctional purine biosynthesis protein PURH, IMPCHASE, phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase, PURHFLJ93545
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ATIC (NP_004035.2).,, Sequence:, MAPGQLALFSVSDKTGLVEFARNLTALGLNLVASGGTAKALRDAGLAVRDVSELTGFPEMLGGRVKTLHPAVHAGILARNIPEDNADMARLDFNLIRVVACNLYPFVKTVASPGVTVEEAVEQIDIGGVTLLRAAAKNHARVTVVCEPEDYVVVSTEMQSSESKDTSLETRRQLALKAFTHTAQYDEAISDYFRKQYSKGVSQMPLRYGMNPHQTPAQLYTLQPKLPITVLNGAPGFINLCDALNAWQLVKELKEALGIPAAASFKHVSPAGAAVGIPLSEDEAKVCMVYDLYKTLTPIS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 471
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.