missing translation for 'onlineSavingsMsg'
Learn More

Collagen XI alpha 2 Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30498031 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498031 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498031

Brand: Novus Biologicals NBP335144AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Collagen XI alpha 2 Polyclonal antibody specifically detects Collagen XI alpha 2 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen XI alpha 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias collagen alpha-2(XI) chain, collagen, type XI, alpha 2, DFNA13, DFNB53, HKE5, PARP, pro-a2 chain of collagen type XI, STL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-380 of human Collagen XI alpha 2 (NP_542411.2).,, Sequence:, IVPGVQAAYESCEQKELECEGGQRERPQNQQPHRAQRSPQQQPSRLHRPQNQEPQSQPTESLYYDYEPPYYDVMTTGTTPDYQDPTPGEEEEILESSLLPPLEEEQTDLQVPPTADRFQAEEYGEGGTDPPEGPYDYTYGYGDDYREETELGPALSAETAHSGAAAHGPRG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1302
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.