missing translation for 'onlineSavingsMsg'
Learn More

UBL3 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30498018 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30498018 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30498018

Brand: Novus Biologicals NBP338069AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBL3 Polyclonal antibody specifically detects UBL3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBL3
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DKFZP434K151, FLJ32018, HCG-1, hsMUB, Membrane-anchored ubiquitin-fold protein, MUB, PNSC1, Protein HCG-1, ubiquitin-like 3, ubiquitin-like protein 3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human UBL3 (NP_009037.1).,, Sequence:, MSSNVPADMINLRLILVSGKTKEFLFSPNDSASDIAKHVYDNWPMDWEEEQVSSPNILRLIYQGRFLHGNVTLGALKLPFGKTTVMHLVARETLPEPNSQGQRNREKTGESNCCVIL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5412
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.