missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ataxin-10 Polyclonal antibody specifically detects Ataxin-10 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Ataxin-10 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ataxin 10, ataxin-10, Brain protein E46 homolog, E46L, FLJ37990, HUMEEP, SCA10spinocerebellar ataxia 10, Spinocerebellar ataxia type 10 protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 346-475 of human Ataxin-10 (NP_037368.1).,, Sequence:, ETTNIFSNCGCVRAEGDISNVANGFKSHLIRLIGNLCYKNKDNQDKVNELDGIPLILDNCNISDSNPFLTQWVIYAIRNLTEDNSQNQDLIAKMEEQGLADASLLKKVGFEVEKKGEKLILKSTRDTPKP |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?