missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7023152
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
705-29-3
C8H6ClF3
194.581
MFCD00000908
XGASTRVQNVVYIZ-UHFFFAOYSA-N
3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride
69716
1-(chloromethyl)-3-(trifluoromethyl)benzene
C1=CC(=CC(=C1)C(F)(F)F)CCl
This item is not returnable. View return policy
705-29-3
C8H6ClF3
194.581
MFCD00000908
XGASTRVQNVVYIZ-UHFFFAOYSA-N
3-trifluoromethyl benzyl chloride, 1-chloromethyl-3-trifluoromethyl benzene, 3-trifluoromethylbenzyl chloride, 3-chloromethyl benzotrifluoride, m-trifluoromethylbenzyl chloride, 3-chloromethyl-benzotrifluoride, benzene, 1-chloromethyl-3-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-m-xylene, 3-chloromethylbenzotrifluoride, 3-trifluoromethylbenzylchloride
69716
1-(chloromethyl)-3-(trifluoromethyl)benzene
C1=CC(=CC(=C1)C(F)(F)F)CCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.254
Boiling Point 68°C to 70°C (12 mmHg)
Flash Point 74°C (165°F)
Odor Pungent
Refractive Index 1.4645
Quantity 5 g
UN Number UN3265
Beilstein 743242
Formula Weight 194.59
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'