missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3522478
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
137386-78-8
C12H9NO2
199.209
MFCD03840110
YGXYNKLOBXMTGL-UHFFFAOYSA-N
3-pyridin-2-yloxy benzaldehyde, 3-pyrid-2-yloxy benzaldehyde, 3-2-pyridinyloxy-benzaldehyde, 3-2-pyridyloxy benzaldehyde, 3-2-pyridinyloxy benzaldehyde
2735363
3-pyridin-2-yloxybenzaldehyde
C1=CC=NC(=C1)OC2=CC=CC(=C2)C=O
This item is not returnable. View return policy
137386-78-8
C12H9NO2
199.209
MFCD03840110
YGXYNKLOBXMTGL-UHFFFAOYSA-N
3-pyridin-2-yloxy benzaldehyde, 3-pyrid-2-yloxy benzaldehyde, 3-2-pyridinyloxy-benzaldehyde, 3-2-pyridyloxy benzaldehyde, 3-2-pyridinyloxy benzaldehyde
2735363
3-pyridin-2-yloxybenzaldehyde
C1=CC=NC(=C1)OC2=CC=CC(=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'