Learn More
3-Pyrid-2-ylbenzaldehyde, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
25 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 183.21 |
Percent Purity | 97% |
Chemical Name or Material | 3-Pyrid-2-ylbenzaldehyde |