missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanone, 95%, Thermo Scientific Chemicals

Catalog Number p-4812970
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6704-31-0
C3H4O2
72.06
ROADCYAOHVSOLQ-UHFFFAOYSA-N
3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n
15024254
oxetan-3-one
C1C(=O)CO1
This item is not returnable. View return policy
6704-31-0
C3H4O2
72.06
ROADCYAOHVSOLQ-UHFFFAOYSA-N
3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n
15024254
oxetan-3-one
C1C(=O)CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 140°C
Flash Point 26°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 72.06
Percent Purity 95%
Chemical Name or Material 3-Oxetanone
compliance-icons compliance-icons
Product Identifier
  • 3-Oxetanone, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'