missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyridazine, 99%, Thermo Scientific Chemicals

Catalog Number p-167506
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1632-76-4
C5H6N2
94.12
MFCD00006469
MXDRPNGTQDRKQM-UHFFFAOYSA-N
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j
74208
3-methylpyridazine
CC1=NN=CC=C1
This item is not returnable. View return policy
1632-76-4
C5H6N2
94.12
MFCD00006469
MXDRPNGTQDRKQM-UHFFFAOYSA-N
pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j
74208
3-methylpyridazine
CC1=NN=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0310g/mL
Boiling Point 214°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5140 to 1.516
Quantity 5 g
Beilstein 23, 92
Specific Gravity 1.031
Formula Weight 94.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methylpyridazine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylpyridazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'