missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylphosphinicopropionic Acid-d3 Sodium Salt, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 3-Methylphosphinicopropionic Acid-d3 Sodium Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | C4 D3 H4 O4 P . 2 Na |
| Quantity | 10 mg |
| Synonym | 3-(Hydroxymethylphosphinyl)propanoic Acid-d3 Sodium Salt, (2-Carboxyethyl)methylphosphinic Acid-d3 Sodium, MPP-d3 Sodium, MPPA-d3 Sodium, Methylphosphinicopropionic Acid-d3 Sodium |
| InChI Formula | InChI=1 S/C4H9O4P0.2 Na/c1-9(7,8)3-2-4(5)6;;/h2-3H2,1H3,(H,5,6)(H,7,8);;/q;2*+1/p-2/i1D3;; |
| SMILES | [Na+].[Na+].[2 H]C([2 H])([2 H])P(=O)([O-])CCC(=O)[O-] |
| IUPAC Name | disodium;3-[oxido(trideuteriomethyl)phosphoryl]propanoate |
| Molecular Weight (g/mol) | 199.068 |
| Formula Weight | 199.0066 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction