missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
A series of novel five-membered urea derivatives as potent NK1 receptor antagonists is described. The effects of substitution of a 4-fluoro group at the phenyl ring and the introduction of an α-methyl group at the benzylic position to improve potency was found. 3,5-Bis(trifluoromethyl)benzyl bromide has been used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
A series of novel five-membered urea derivatives as potent NK1 receptor antagonists is described. The effects of substitution of a 4-fluoro group at the phenyl ring and the introduction of an α-methyl group at the benzylic position to improve potency was found. 3,5-Bis(trifluoromethyl)benzyl bromide has been used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
Specifications
Quantity | 250 mg |
UN Number | UN3265 |
Sensitivity | Moisture sensitive |
Formula Weight | 253.06 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methyl-5-(trifluoromethyl)benzyl bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'