missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals

Catalog Number p-7033520
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3347-62-4
C10H10N2
158.20
MFCD00022385,MFCD00524752
QHRSESMSOJZMCO-UHFFFAOYSA-N
18774
5-methyl-3-phenyl-1H-pyrazole
CC1=CC(=NN1)C1=CC=CC=C1
This item is not returnable. View return policy
3347-62-4
C10H10N2
158.20
MFCD00022385,MFCD00524752
QHRSESMSOJZMCO-UHFFFAOYSA-N
18774
5-methyl-3-phenyl-1H-pyrazole
CC1=CC(=NN1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 124°C to 127°C
Quantity 1 g
Synonym 3-methyl-5-phenyl-1h-pyrazole, femerazol, 3-methyl-5-phenylpyrazole, phemerazole, 1h-pyrazole, 3-methyl-5-phenyl, 5-methyl-3-phenylpyrazole, 1h-pyrazole,3-methyl-5-phenyl, pyrazole, 3-methyl-5-phenyl, unii-0xfx19d77a, 1h-pyrazole, 5-methyl-3-phenyl
Formula Weight 158.2
Chemical Name or Material 3-Methyl-5-phenyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'