Learn More
3-Methoxypropionic acid, 97+%, Thermo Scientific Chemicals
Catalog Number p-7077416
Click to view available options
Quantity:
10 mL
50 mL
2544-06-1
C4H8O3
104.11
YSIKHBWUBSFBRZ-UHFFFAOYSA-N
134442
3-methoxypropanoic acid
COCCC(=O)O
2544-06-1
C4H8O3
104.11
YSIKHBWUBSFBRZ-UHFFFAOYSA-N
134442
3-methoxypropanoic acid
COCCC(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.1080g/mL
Boiling Point 116.0°C (9.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 10 mL
Specific Gravity 1.108
Synonym 3-methoxypropionic acid, propanoic acid, 3-methoxy, propanoic acid, methoxy, m-peg1-acid, propanoic acid,methoxy, acmc-1aiwk, 3-methoxy-propionic acid, #, propanoic acid,3-methoxy, .beta.-methoxypropionic acid
Formula Weight 104.11
Percent Purity 97+%
Chemical Name or Material 3-Methoxypropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxypropionic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.