missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7043682
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
5071-96-5
C8H11NO
137.18
MFCD00008115
GRRIMVWABNHKBX-UHFFFAOYSA-N
3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
21156
(3-methoxyphenyl)methanamine
COC1=CC=CC(CN)=C1
This item is not returnable. View return policy
5071-96-5
C8H11NO
137.18
MFCD00008115
GRRIMVWABNHKBX-UHFFFAOYSA-N
3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
21156
(3-methoxyphenyl)methanamine
COC1=CC=CC(CN)=C1

3-Methoxybenzylamine is used in the preparation of 6-substituted purines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzylamine is used in the preparation of 6-substituted purines

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 1.066
Boiling Point 230°C to 231°C
Flash Point >110°C (230°F)
Refractive Index 1.547
Quantity 5 g
UN Number UN2735
Beilstein 1447182
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'