missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Catalog Number p-7052424
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1214328-36-5
C9H8F3NO2
219.163
MFCD13185679
ICTJARUSYAWEDK-UHFFFAOYSA-N
3-methoxy-4-trifluoromethyl benzamide
66523618
3-methoxy-4-(trifluoromethyl)benzamide
COC1=C(C=CC(=C1)C(=O)N)C(F)(F)F
This item is not returnable. View return policy
1214328-36-5
C9H8F3NO2
219.163
MFCD13185679
ICTJARUSYAWEDK-UHFFFAOYSA-N
3-methoxy-4-trifluoromethyl benzamide
66523618
3-methoxy-4-(trifluoromethyl)benzamide
COC1=C(C=CC(=C1)C(=O)N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 219°C to 220°C
Color White
Quantity 1 g
Formula Weight 219.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-4-(trifluoromethyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'